Thermo Scientific Chemicals

(R)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H63991.14
25 g, Each
Thermo Scientific Chemicals

(R)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H63991.14
25 g, Each
Quantity
Catalog number: H63991.14
also known as H63991-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
45121-22-0
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Molecular Formula
C9H17NO4
InChI Key
PNFVIPIQXAIUAY-UHFFFAOYNA-N
SMILES
CCC(NC(=O)OC(C)(C)C)C(O)=O
Assay (HPLC)
>94.0%

Description

(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

Solubility
Soluble in DMF (1mmol in 1ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text