N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H63993.06
also known as H63993-06
Price (USD)/ Each
298.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
298.00
Add to cart
N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%, Thermo Scientific Chemicals
Catalog numberH63993.06
Price (USD)/ Each
298.00
-
Add to cart
Chemical Identifiers
CAS64187-48-0
IUPAC Name1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular FormulaC14H17NO5
InChI KeyVVKAGQHUUDRPOI-UHFFFAOYNA-N
SMILESCOC(=O)C1CC(O)CN1C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only