Thermo Scientific Chemicals

1-Methyl-2-imidazolemethanol, 98%, Thermo Scientific Chemicals

Catalog number: H64006.06
5 g, Each
Thermo Scientific Chemicals

1-Methyl-2-imidazolemethanol, 98%, Thermo Scientific Chemicals

Catalog number: H64006.06
5 g, Each
Quantity
Catalog number: H64006.06
also known as H64006-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17334-08-6
IUPAC Name
(1-methyl-1H-imidazol-2-yl)methanol
Molecular Formula
C5H8N2O
InChI Key
CDQDMLWGTVLQEE-UHFFFAOYSA-N
SMILES
CN1C=CN=C1CO
Assay (HPLC)
>97.5%

Description

1-Methyl-2-imidazolemethanol is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-imidazolemethanol is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid, acid chlorides, halogens, halocarbons.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text