Thermo Scientific Chemicals

Methyl 6-chloropyridine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H64023.06
5 g, Each
Thermo Scientific Chemicals

Methyl 6-chloropyridine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H64023.06
5 g, Each
Quantity
Catalog number: H64023.06
also known as H64023-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6636-55-1
IUPAC Name
methyl 6-chloropyridine-2-carboxylate
Molecular Formula
C7H6ClNO2
InChI Key
TWUXBVMXSBEKHA-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(Cl)=N1
Assay (HPLC)
>94.0%

Description

Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2?-bipyridine-6-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2′-bipyridine-6-carboxylate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text