Thermo Scientific Chemicals

Thiazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H64028.06
5 g, Each
Thermo Scientific Chemicals

Thiazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H64028.06
5 g, Each
Quantity
Catalog number: H64028.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3364-80-5
IUPAC Name
1,3-thiazole-4-carbaldehyde
Molecular Formula
C4H3NOS
InChI Key
WRFKSVINLIQRKF-UHFFFAOYSA-N
SMILES
O=CC1=CSC=N1
Assay (HPLC)
>94.0%

Description

Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text