Thermo Scientific Chemicals

4-Bromo-2,3-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64044.03
1 g, Each
Thermo Scientific Chemicals

4-Bromo-2,3-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64044.03
1 g, Each
Quantity
Catalog number: H64044.03
also known as H64044-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
644985-24-0
IUPAC Name
4-bromo-2,3-difluorobenzaldehyde
Molecular Formula
C7H3BrF2O
InChI Key
IMOLPSNRLZLWQR-UHFFFAOYSA-N
SMILES
FC1=C(Br)C=CC(C=O)=C1F
Form
Powder
Appearance (Color)
Off-white
Assay (HPLC)
>97.5%

Description

4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text