2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals
2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number H64066.14
also known as H64066-14
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals
Catalog numberH64066.14
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS22536-67-0
IUPAC Name2,5-dichloropyrimidine
Molecular FormulaC4H2Cl2N2
InChI KeyCEJAHXLRNZJPQH-UHFFFAOYSA-N
SMILESClC1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Alexander J. Blake; Paul T. Brain; Hamish McNab; Jennifer Miller; Carole A. Morrison; Simon Parsons; David W. H. Rankin; Heather E. Robertson; and Bruce A. Smart. Structure Analysis Restrained by ab Initio Calculations: The Molecular Structure of 2,5-Dichloropyrimidine in Gaseous and Crystalline Phases. J. Phys. Chem.1996100(30), 12280-12287.
  2. Donald G. Crosby; Robert V. Berthold. n-Butyl 5-Chloro-2-pyrimidoxyacetate—A Plant Growth Regulator Analog. J. Org. Chem.196025(11), 1916-1919.