Thermo Scientific Chemicals

2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals

Catalog number: H64066.22
100 g, Each
Thermo Scientific Chemicals

2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals

Catalog number: H64066.22
100 g, Each
Quantity
Catalog number: H64066.22
also known as H64066-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22536-67-0
IUPAC Name
2,5-dichloropyrimidine
Molecular Formula
C4H2Cl2N2
InChI Key
CEJAHXLRNZJPQH-UHFFFAOYSA-N
SMILES
ClC1=CN=C(Cl)N=C1
Assay (HPLC)
>94.0%

Description

2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text