Thermo Scientific Chemicals

2-Hydrazinopyrazine, 98%, Thermo Scientific Chemicals

Catalog number: H64086.06
5 g, Each
Thermo Scientific Chemicals

2-Hydrazinopyrazine, 98%, Thermo Scientific Chemicals

Catalog number: H64086.06
5 g, Each
Quantity
Catalog number: H64086.06
also known as H64086-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54608-52-5
IUPAC Name
2-hydrazinylpyrazine
Molecular Formula
C4H6N4
InChI Key
IVRLZJDPKUSDCF-UHFFFAOYSA-N
SMILES
NNC1=NC=CN=C1
Assay (HPLC)
>97.5%

Description

Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text