Thermo Scientific Chemicals

Ethyl imidazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H64095.14
25 g, Each
Thermo Scientific Chemicals

Ethyl imidazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H64095.14
25 g, Each
Quantity
Catalog number: H64095.14
also known as H64095-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23785-21-9
IUPAC Name
ethyl 1H-imidazole-5-carboxylate
Molecular Formula
C6H8N2O2
InChI Key
KLWYPRNPRNPORS-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CN=CN1
Assay (HPLC)
>97.5%

Description

A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text