Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H64102.06
also known as H64102-06
Price (USD)/ Each
294.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
346.00 
Save 51.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
294.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
346.00 
Save 51.35 (15%)
Add to cart
Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH64102.06
Price (USD)/ Each
294.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
346.00 
Save 51.35 (15%)
-
Add to cart
Chemical Identifiers
CAS71233-25-5
IUPAC Name1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate
Molecular FormulaC13H21NO5
InChI KeyWCTXJAXKORIYNA-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only