Thermo Scientific Chemicals

1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: H64116.14
25 g, Each
Thermo Scientific Chemicals

1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: H64116.14
25 g, Each
Quantity
Catalog number: H64116.14
also known as H64116-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3163-27-7
IUPAC Name
1-(bromomethyl)naphthalene
Molecular Formula
C11H9Br
InChI Key
RZJGKPNCYQZFGR-UHFFFAOYSA-N
SMILES
BrCC1=C2C=CC=CC2=CC=C1
Assay (GC)
≥94.0%

Description

It is employed as a intermediate for organic synthesis and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text