Thermo Scientific Chemicals

7-Bromo-1-heptene, 97%, Thermo Scientific Chemicals

Catalog number: H64127.14
25 g, Each
Thermo Scientific Chemicals

7-Bromo-1-heptene, 97%, Thermo Scientific Chemicals

Catalog number: H64127.14
25 g, Each
Quantity
Catalog number: H64127.14
also known as H64127-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4117-09-3
IUPAC Name
7-bromohept-1-ene
Molecular Formula
C7H13Br
InChI Key
GNYDYUQVALBGGZ-UHFFFAOYSA-N
SMILES
BrCCCCCC=C
Form
Liquid
Appearance (Color)
Colorless to pale yellow
Assay (GC)
>96.0%

Description

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text