Thermo Scientific Chemicals

4-Bromophthalimide, ≥97%, Thermo Scientific Chemicals

Catalog number: H64136.06
5 g, Each
Thermo Scientific Chemicals

4-Bromophthalimide, ≥97%, Thermo Scientific Chemicals

Catalog number: H64136.06
5 g, Each
Quantity
Catalog number: H64136.06
also known as H64136-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6941-75-9
IUPAC Name
5-bromo-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C8H4BrNO2
InChI Key
GNYICZVGHULCHE-UHFFFAOYSA-N
SMILES
BrC1=CC=C2C(=O)NC(=O)C2=C1
Assay (HPLC)
>97.0%

Description

It is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Soluble in N,N-DMF almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text