Thermo Scientific Chemicals

3-Amino-1-adamantanol, ≥98%, Thermo Scientific Chemicals

Catalog number: H64137.14
25 g, Each
Thermo Scientific Chemicals

3-Amino-1-adamantanol, ≥98%, Thermo Scientific Chemicals

Catalog number: H64137.14
25 g, Each
Quantity
Catalog number: H64137.14
also known as H64137-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
702-82-9
IUPAC Name
3-aminoadamantan-1-ol
Molecular Formula
C10H17NO
InChI Key
DWPIPTNBOVJYAD-UHFFFAOYNA-N
SMILES
NC12CC3CC(C1)CC(O)(C3)C2
Assay (GC)
>98.0%

Description

It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

Solubility
Soluble in organic solvents,insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text