Thermo Scientific Chemicals

2,5-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H64140.06
5 g, Each
Thermo Scientific Chemicals

2,5-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H64140.06
5 g, Each
Quantity
Catalog number: H64140.06
also known as H64140-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4925-88-6
IUPAC Name
2,5-dimethoxy-4-methylbenzaldehyde
Molecular Formula
C10H12O3
InChI Key
LRSRTWLEJBIAIT-UHFFFAOYSA-N
SMILES
COC1=CC(C)=C(OC)C=C1C=O
Assay (HPLC)
>96.0%

Description

Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text