2-Amino-5-(trifluoromethyl)benzoic acid, 95%, Thermo Scientific Chemicals
2-Amino-5-(trifluoromethyl)benzoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-(trifluoromethyl)benzoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H64141.MD
also known as H64141-MD
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
2-Amino-5-(trifluoromethyl)benzoic acid, 95%, Thermo Scientific Chemicals
Catalog numberH64141.MD
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS83265-53-6
IUPAC Name2-amino-5-(trifluoromethyl)benzoic acid
Molecular FormulaC8H6F3NO2
InChI KeyGLCQUPLYYXSPQB-UHFFFAOYSA-N
SMILESNC1=C(C=C(C=C1)C(F)(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only