Used for proteomics research. Also employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used for proteomics research. Also employed as a intermediate for pharmaceutical.
Solubility
Soluble in chloroform and tetrahydrofuran (THF).
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
Xin Wu; Mufaro A. Dube; Albert J. Fry. Electrophilic nitration of triphenylamines as a route to high oxidation potential electrocatalysts. Polynitration, nitrodebromination, and bromine dance. Tetrahedron Letters. 200647(44), 7667-7669.