Thermo Scientific Chemicals

4,4',4''-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals

Catalog number: H64162.14
25 g, Each
Thermo Scientific Chemicals

4,4',4''-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals

Catalog number: H64162.14
25 g, Each
Quantity
Catalog number: H64162.14
also known as H64162-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1159-53-1
IUPAC Name
4-methyl-N,N-bis(4-methylphenyl)aniline
Molecular Formula
C21H21N
InChI Key
YXYUIABODWXVIK-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Assay (HPLC)
>97.5%

Description

Used for proteomics research. Also employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for proteomics research. Also employed as a intermediate for pharmaceutical.

Solubility
Soluble in chloroform and tetrahydrofuran (THF).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text