Thermo Scientific Chemicals

2,3,5-Trichloropyridine, ≥98%, Thermo Scientific Chemicals

Catalog number: H64173.22
100 g, Each
Thermo Scientific Chemicals

2,3,5-Trichloropyridine, ≥98%, Thermo Scientific Chemicals

Catalog number: H64173.22
100 g, Each
Quantity
Catalog number: H64173.22
also known as H64173-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16063-70-0
IUPAC Name
2,3,5-trichloropyridine
Molecular Formula
C5H2Cl3N
InChI Key
CNLIIAKAAMFCJG-UHFFFAOYSA-N
SMILES
ClC1=CN=C(Cl)C(Cl)=C1
Assay (HPLC)
>98.0%

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text