3,5-Dibromo-4-methoxypyridine, 98%, Thermo Scientific Chemicals
3,5-Dibromo-4-methoxypyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dibromo-4-methoxypyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H64177.06
also known as H64177-06
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
3,5-Dibromo-4-methoxypyridine, 98%, Thermo Scientific Chemicals
Catalog numberH64177.06
Price (USD)/ Each
276.00
-
Add to cart
Chemical Identifiers
CAS650140-84-4
IUPAC Name3,5-dibromo-4-methoxypyridin-1-ium-1-olate
Molecular FormulaC6H5Br2NO2
InChI KeyKAYQEYHOGFRTAL-UHFFFAOYSA-N
SMILESCOC1=C(Br)C=[N+]([O-])C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only