Thermo Scientific Chemicals

1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog number: H64191.06
5 g, Each
Thermo Scientific Chemicals

1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog number: H64191.06
5 g, Each
Quantity
Catalog number: H64191.06
also known as H64191-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4965-09-7
IUPAC Name
1-methyl-1,2,3,4-tetrahydroisoquinoline
Molecular Formula
C10H13N
InChI Key
QPILYVQSKNWRDD-UHFFFAOYNA-N
SMILES
CC1NCCC2=CC=CC=C12
Assay (GC)
>94.0%

Description

1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity. Following systemic administration in rats, 1-Methyl-1,2,3,4-tetrahydroisoquinoline produces antidepressant-like effect similar to the effect of imipramine. 1MeTIQ is a reversible short-acting moderate inhibitor of MAO A/B. Also known as 1,2,3,4-Tetrahydro-1-methylisoquinoline, 1MeTIQ. It is an important raw material and intermediate for revaprazan and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text