4-Formylbenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
4-Formylbenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Formylbenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H64192.06
also known as H64192-06
Price (USD)/ Each
86.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
86.60
Add to cart
4-Formylbenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Catalog numberH64192.06
Price (USD)/ Each
86.60
-
Add to cart
Chemical Identifiers
CAS128376-64-7
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular FormulaC13H17BO3
InChI KeyDMBMXJJGPXADPO-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point54-61?C
FormPowder
Appearance (Color)White to cream
Assay from Supplier's CofA≥97.5%
A boronic acid benzaldehyde compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A boronic acid benzaldehyde compound for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Alexander Oehlke; Alexander A. Auer; Ina Jahre; Bernhard Walfort; Tobias Rüffer; Petra Zoufalá; Heinrich Lang; and Stefan Spange. Nitro-Substituted Stilbeneboronate Pinacol Esters and Their Fluoro-Adducts. Fluoride Ion Induced Polarity Enhancement of Arylboronate Esters. J. Org. Chem. 200772(12), 4328-4339.
  2. Yiting Wang; Jinchong Xiaoa; Sujuan Wang; Bo Yang; & Xinwu Ba. Tunable fluorescent sensing of cysteine and homocysteine by intramolecular charge transfer. Supramolecular Chemistry. 201022(6), 380-386.