Thermo Scientific Chemicals

2-Fluoro-5-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64210.14
25 g, Each
Thermo Scientific Chemicals

2-Fluoro-5-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64210.14
25 g, Each
Quantity
Catalog number: H64210.14
also known as H64210-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
456-24-6
IUPAC Name
2-fluoro-5-nitropyridine
Molecular Formula
C5H3FN2O2
InChI Key
XOZAJNLUAODXSP-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)N=C1
Assay (GC)
>97.5%

Description

It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text