2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64238.03
also known as H64238-03
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
Add to cart
2-Chloro-4-bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberH64238.03
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
-
Add to cart
Chemical Identifiers
CAS1214386-29-4
IUPAC Name4-bromo-2-chloro-5-fluorobenzaldehyde
Molecular FormulaC7H3BrClFO
InChI KeyXPEGZWLIWARWAA-UHFFFAOYSA-N
SMILESFC1=C(Br)C=C(Cl)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only