Thermo Scientific Chemicals

4-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64244.03
1 g, Each
Thermo Scientific Chemicals

4-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64244.03
1 g, Each
Quantity
Catalog number: H64244.03
also known as H64244-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30766-03-1
IUPAC Name
4-bromopyridine-2-carboxylate
Molecular Formula
C6H3BrNO2
InChI Key
RPHHYRNGCJYQSP-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC(Br)=CC=N1
Assay (HPLC)
>96.0%

Description

4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text