5-Bromo-2-hydroxy-3-methylpyridine, 98%, Thermo Scientific Chemicals
5-Bromo-2-hydroxy-3-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-hydroxy-3-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number H64253.22
also known as H64253-22
Price (USD)/ Each
506.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
506.00
Add to cart
5-Bromo-2-hydroxy-3-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberH64253.22
Price (USD)/ Each
506.00
-
Add to cart
Chemical Identifiers
CAS89488-30-2
IUPAC Name5-bromo-3-methyl-1,2-dihydropyridin-2-one
Molecular FormulaC6H6BrNO
InChI KeyHXPMRPRBABWPKL-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only