It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Dušan Z. Mijin; Mostafa Baghbanzadeh; Claudia Reidlinger; C. Oliver Kappe. The microwave-assisted synthesis of 5-arylazo-4,6-disubstituted-3-cyano-2-pyridone dyes. Dyes and Pigments. 2010 85(1-2), 73-78.
- Sunil S. Chavan; Mariam S. Degani. Ionic Liquid Catalyzed 4,6-Disubstituted-3-Cyano-2-Pyridone Synthesis Under Solvent-Free Conditions. Catalysis Letters. 2011 141(11), 1693-1697.