Thermo Scientific Chemicals

Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals

Catalog number: H64255.22
100 g, Each
Thermo Scientific Chemicals

Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific Chemicals

Catalog number: H64255.22
100 g, Each
Quantity
Catalog number: H64255.22
also known as H64255-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5597-50-2
IUPAC Name
methyl 3-(4-hydroxyphenyl)propanoate
Molecular Formula
C10H12O3
InChI Key
XRAMJHXWXCMGJM-UHFFFAOYSA-N
SMILES
COC(=O)CCC1=CC=C(O)C=C1
Assay (HPLC)
>97.5%

Description

Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text