Chemical Identifiers
CAS1463-10-1
IUPAC Name1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H14N2O6
InChI KeyDWRXFEITVBNRMK-JXOAFFINSA-N
SMILESCC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)NC1=O
View more