Thermo Scientific Chemicals

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64288.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-amidinoacetate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64288.14
25 g, Each
Quantity
Catalog number: H64288.14
also known as H64288-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57508-48-2
IUPAC Name
(1-amino-3-ethoxy-3-oxopropylidene)azanium
Molecular Formula
C5H11N2O2
InChI Key
HSDKTLKBDJXJQU-UHFFFAOYSA-O
SMILES
CCOC(=O)CC(N)=[NH2+]
Assay (HPLC)
>94.0%

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text