Thermo Scientific Chemicals

Glycidyl (S)-(+)-3-nitrobenzenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: H64289.14
25 g, Each
Thermo Scientific Chemicals

Glycidyl (S)-(+)-3-nitrobenzenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: H64289.14
25 g, Each
Quantity
Catalog number: H64289.14
also known as H64289-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115314-14-2
IUPAC Name
[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Molecular Formula
C9H9NO6S
InChI Key
AIHIHVZYAAMDPM-QMMMGPOBSA-N
SMILES
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
Assay (GC)
>97.5%

Description

Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

Solubility
Soluble in chloroform, methylene chloride and tetrahydrofuran. Insoluble in hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text