4-Methoxy-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals
4-Methoxy-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64295.03
also known as H64295-03
Price (USD)/ Each
44.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
44.70
Add to cart
4-Methoxy-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals
Catalog numberH64295.03
Price (USD)/ Each
44.70
-
Add to cart
Chemical Identifiers
CAS52289-54-0
IUPAC Name4-methoxy-2-methylbenzaldehyde
Molecular FormulaC9H10O2
InChI KeyWICYVKGMEJSDAO-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only