3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water (9.8 g/L).
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- A. R. Pinder. Pyrrolidine, piperidine, and pyridine alkaloids. Nat. Prod. Rep. 19896 (5), 515-521.
- Amy S. Manoso; Chuljin Ahn; Arash Soheili; Christopher J. Handy; Reuben Correia; W. Michael Seganish; and Philip DeShong . Improved Synthesis of Aryltrialkoxysilanes via Treatment of Aryl Grignard or Lithium Reagents with Tetraalkyl Orthosilicates. J. Org. Chem. 198969 (24), 8305-8314.