6-Bromo-2-methyl-3-nitropyridine, 97%, Thermo Scientific Chemicals
6-Bromo-2-methyl-3-nitropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromo-2-methyl-3-nitropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H64299.14
also known as H64299-14
Price (USD)/ Each
154.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
154.00
Add to cart
6-Bromo-2-methyl-3-nitropyridine, 97%, Thermo Scientific Chemicals
Catalog numberH64299.14
Price (USD)/ Each
154.00
-
Add to cart
Chemical Identifiers
CAS22282-96-8
IUPAC Name6-bromo-2-methyl-3-nitropyridine
Molecular FormulaC6H5BrN2O2
InChI KeyPCNVKBBKROTUNS-UHFFFAOYSA-N
SMILESCC1=C(C=CC(Br)=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only