Thermo Scientific Chemicals

1-Bromo-4-fluorobutane, 97%, Thermo Scientific Chemicals

Catalog number: H64317.03
1 g, Each
Thermo Scientific Chemicals

1-Bromo-4-fluorobutane, 97%, Thermo Scientific Chemicals

Catalog number: H64317.03
1 g, Each
Quantity
Catalog number: H64317.03
also known as H64317-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
462-72-6
IUPAC Name
1-bromo-4-fluorobutane
Molecular Formula
C4H8BrF
InChI Key
WMCUHRDQSHQNRW-UHFFFAOYSA-N
SMILES
FCCCCBr
Assay (GC)
>96.0%

Description

1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

Solubility
Sparingly soluble in water at 25°C 0.48 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text