Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals
Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64327.03
also known as H64327-03
Price (USD)/ Each
37.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
37.50
Add to cart
Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals
Catalog numberH64327.03
Price (USD)/ Each
37.50
-
Add to cart
Chemical Identifiers
CAS29681-42-3
IUPAC Namemethyl 4-bromopyridine-2-carboxylate
Molecular FormulaC7H6BrNO2
InChI KeyJZFLATQBIPILFS-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA>97.0%
Infrared spectrumConforms
Appearance (Color)Light cream to cream
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only
Tristan Verdelet, et al. Straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals.Tetrahedron.,2011,67(45), 8757-8762.