Thermo Scientific Chemicals

Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals

Catalog number: H64327.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals

Catalog number: H64327.14
25 g, Each
Quantity
Catalog number: H64327.14
also known as H64327-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29681-42-3
IUPAC Name
methyl 4-bromopyridine-2-carboxylate
Molecular Formula
C7H6BrNO2
InChI Key
JZFLATQBIPILFS-UHFFFAOYSA-N
SMILES
COC(=O)C1=NC=CC(Br)=C1
Form
Powder
Assay from Supplier's CofA
>97.0%
Infrared spectrum
Conforms
Appearance (Color)
Light cream to cream

Description

It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text