Thermo Scientific Chemicals

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog number: H64329.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog number: H64329.06
5 g, Each
Quantity
Catalog number: H64329.06
also known as H64329-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
887707-25-7
Assay (HPLC)
>94.0%

Description

It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text