Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64340.06
also known as H64340-06
Price (USD)/ Each
51.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
51.50
Add to cart
Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Catalog numberH64340.06
Price (USD)/ Each
51.50
-
Add to cart
Chemical Identifiers
CAS5337-03-1
IUPAC Nameoxane-4-carboxylic acid
Molecular FormulaC6H10O3
InChI KeyAVPKHOTUOHDTLW-UHFFFAOYSA-N
SMILESOC(=O)C1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥97.5%
FormCrystalline powder
Melting Point87-89?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only