Thermo Scientific Chemicals

Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H64340.14
25 g, Each
Thermo Scientific Chemicals

Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H64340.14
25 g, Each
Quantity
Catalog number: H64340.14
also known as H64340-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5337-03-1
IUPAC Name
oxane-4-carboxylic acid
Molecular Formula
C6H10O3
InChI Key
AVPKHOTUOHDTLW-UHFFFAOYSA-N
SMILES
OC(=O)C1CCOCC1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥97.5%
Form
Crystalline powder
Melting Point
87-89?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text