Thermo Scientific Chemicals

beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64346.06
5 g, Each
Thermo Scientific Chemicals

beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64346.06
5 g, Each
Quantity
Catalog number: H64346.06
also known as H64346-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58620-93-2
IUPAC Name
tert-butyl 3-aminopropanoate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
DOMTZTVJNZKUNX-UHFFFAOYSA-N
SMILES
Cl.CC(C)(C)OC(=O)CCN
Assay (Titration)
>94.0% to <106.0%

Description

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text