beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals
beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number H64346.14
also known as H64346-14
Price (USD)/ Each
229.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
229.00
Add to cart
beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH64346.14
Price (USD)/ Each
229.00
-
Add to cart
Chemical Identifiers
CAS58620-93-2
IUPAC Nametert-butyl 3-aminopropanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyDOMTZTVJNZKUNX-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>94.0% to <106.0%
Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Nigel I Howard; Timothy D.H Bugg. Synthesis and activity of 5'-Uridinyl dipeptide analogues mimicking the amino terminal peptide chain of nucleoside antibiotic mureidomycin A. Journal of Heterocyclic Chemistry. 200311 (14), 3083-3099.
  2. Shigeru Kuwata;Akihiro Nakanishi; Takashi Yamada; Toshifumi Miyazawa. Total synthesis of Leucinostatin D. Tetrahedron Letters. 200333 (46), 6995-6998.