Thermo Scientific Chemicals

2-Methylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64375.03
1 g, Each
Thermo Scientific Chemicals

2-Methylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64375.03
1 g, Each
Quantity
Catalog number: H64375.03
also known as H64375-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5194-32-1
IUPAC Name
2-methylpyrimidine-5-carboxylic acid
Molecular Formula
C6H6N2O2
InChI Key
NMGIXZFBQPETOK-UHFFFAOYSA-N
SMILES
CC1=NC=C(C=N1)C(O)=O
Proton NMR
>96.0%

Description

It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store in freezer at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text