L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals
L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H64384.14
Price (USD)/ Each
465.65
Online exclusive
517.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
465.65
Online exclusive
517.00 
Save 51.35 (10%)
Add to cart
L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals
Catalog numberH64384.14
Price (USD)/ Each
465.65
Online exclusive
517.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS45172-15-4
IUPAC Nameethyl (2S)-2,6-diisocyanatohexanoate
Molecular FormulaC10H14N2O4
InChI KeyIGWTZHMYJZZIGC-IMWMWJONNA-N
SMILESCCOC(=O)[C@H](CCCCN=C=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Assay (HPLC)?96.0%
Appearance (Color)Yellow
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water (0.19 g/L).

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Jian Han; Bing Chen; Lin Ye; Ai-ying Zhang; Jian Zhang; Zeng-guo Feng. Synthesis and characterization of biodegradable polyurethane based on poly(ε-caprolactone) and L-lysine ethyl ester diisocyanate. Frontiers of Materials Science in China. 20093 (1), 25-32.
  2. R. F. Storey; J. S. Wiggins; and A. D. Puckett. Hydrolyzable poly(ester-urethane) networks from L-lysine diisocyanate and D,L-lactide/ϵ-caprolactone homo- and copolyester triols. Journal of Polymer Science Part A: Polymer Chemistry. 199432 (12), 2345-2363.