Thermo Scientific Chemicals

Methyl 2-amino-4,5-difluorobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H64385.22
100 g, Each
Thermo Scientific Chemicals

Methyl 2-amino-4,5-difluorobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H64385.22
100 g, Each
Quantity
Catalog number: H64385.22
also known as H64385-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207346-42-7
IUPAC Name
methyl 2-amino-4,5-difluorobenzoate
Molecular Formula
C8H7F2NO2
InChI Key
VIAQNTRKUPBQKR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(F)=C(F)C=C1N
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps
Assay from Supplier's CofA
≥97.5% (GC)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text