Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals
Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H64389.MD
also known as H64389-MD
Price (USD)/ Each
78.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
78.80
Add to cart
Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals
Catalog numberH64389.MD
Price (USD)/ Each
78.80
-
Add to cart
Chemical Identifiers
CAS317830-83-4
IUPAC Name(1-benzofuran-3-yl)boronic acid
Molecular FormulaC8H7BO3
InChI KeyDFUGYZQSDFQVPU-UHFFFAOYSA-N
SMILESOB(O)C1=COC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only