Thermo Scientific Chemicals

Methyl 6-bromo-5-fluoronicotinate, 98%, Thermo Scientific Chemicals

Catalog number: H64395.06
5 g, Each
Thermo Scientific Chemicals

Methyl 6-bromo-5-fluoronicotinate, 98%, Thermo Scientific Chemicals

Catalog number: H64395.06
5 g, Each
Quantity
Catalog number: H64395.06
also known as H64395-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1214336-88-5
IUPAC Name
methyl 6-bromo-5-fluoropyridine-3-carboxylate
Molecular Formula
C7H5BrFNO2
InChI Key
PXHKAVXJQLVUBI-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(F)=C(Br)N=C1
Assay (HPLC)
>97.5%

Description

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent. Store in freezer at (-20°C).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text