Thermo Scientific Chemicals

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: H64396.06
5 g, Each
Thermo Scientific Chemicals

(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: H64396.06
5 g, Each
Quantity
Catalog number: H64396.06
also known as H64396-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87583-89-9
IUPAC Name
N1,N2-dimethylcyclohexane-1,2-diamine
Molecular Formula
C8H18N2
InChI Key
JRHPOFJADXHYBR-UHFFFAOYNA-N
SMILES
CNC1CCCCC1NC
Assay (GC)
>97.5%

Description

It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 6-substituted 1-oxoindanoyl isoleucine conjugates and modeling studies with the coi1-jaz co-receptor complex of lima bean.

Solubility
Slightly soluble in water.

Notes
Store away from acids, air, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text