Thermo Scientific Chemicals

Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H64417.06
5 g, Each
Thermo Scientific Chemicals

Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H64417.06
5 g, Each
Quantity
Catalog number: H64417.06
also known as H64417-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50593-91-4
IUPAC Name
methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Molecular Formula
C7H7BrN2O2S
InChI Key
MYZJIEWTRJTWCD-UHFFFAOYSA-N
SMILES
COC(=O)C1=NC(SC)=NC=C1Br
Assay (HPLC)
>96.0%

Description

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water (0.42 g/L).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text